missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TGFB2 Recombinant Human Protein

Product Code. p-5232847
Click to view available options
Quantity:
5 μg
Unit Size:
5µg
Each
This item is not returnable. View return policy

Product Code. 10272593

Brand: Gibco™ PHG9114

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Human Transforming Growth Factor β 2 (TGFB2) is a bioactive protein intended for use in cell culture applications

TGFB2 binds the transforming growth factor-β receptor and plays a role in regulation of cell growth and proliferation. It may be involved in mesenchymal-epithelial cell interactions during development.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by its ability to inhibit the mouse IL-4 dependent proliferation of mouse HT-2 cells

Approved for shipment on Wet or Dry Ice.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Gene ID (Entrez) 7042
Molecular Weight (g/mol) 25 kDa
Purification Method Sequential Chromatography
Quantity 5 μg
Gene Alias TGF-β2
Species Human
Recombinant Recombinant
Content And Storage Store in freezer (-5 to -30°C).
Endotoxin Level < 0.1 ng/μg
Mass 5 μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System Baculovirus
Protein Family Growth Factors & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype TGFs (Transforming Growth Factors)
Research Category Immunology, Neurobiology, Oncology, Stem Cell Research, Cardiovascular Research, Differentiation, Bone Research
Product Line Gibco
Purity or Quality Grade 98 %
Show More Show Less

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.