missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CD40 Ligand (TNFSF5) Recombinant Protein

Product Code. 10494553
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10494553

Brand: Gibco™ PHP0025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute sCD40L in 5-10 mM sodium phosphate, pH 7.5, to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: The lyophilized protein is stable at room temperature for 1 month, but should be kept at -20°C for long term storage, preferably desiccated. Working aliquots stored with a carrier protein are stable for at least 3 months at -20°C to -80°C. Avoid repeated freeze/thaw cycles.

CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29965
For Use With (Application) Bioactivity
Formulation 10mM sodium phosphate with no preservative
Gene ID (Entrez) 959
Molecular Weight (g/mol) 16.3 kDa
Name Human CD40 Ligand (TNFSF5)
Purification Method Purified
Quantity 50 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/μg
Gene Alias CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5
Common Name CD154 (CD40 Ligand)
Gene Symbol CD40LG
Biological Activity ED50 = 5 - 10 ng/mL; determined by the dose-dependent stimulation of IL-8 production by human PBMCs.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence Human CD40 ligand recombinant protein contains 149 amino acids
Content And Storage -20°C
Activity ED50 = 5 - 10 ng/mL; determined by the dose-dependent stimulation of IL-8 production by human PBMCs.
Endotoxin Level <0.1 ng/μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Cytokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype Other Proteins
Research Category Immunology, Stem Cell Research, Signal Transduction
Product Line Gibco
Purity or Quality Grade ≥98% by SDS-PAGE and HPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.