missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Sf21 cells in Sf-900™ II SFM

Product Code. 10686103
Change view
Click to view available options
Quantity:
1.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10686103 1.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Review the Material Safety Data Sheet before handling.
Product Code. 10686103 Supplier Gibco™ Supplier No. 11497013
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Review the Material Safety Data Sheet before handling.

Used to isolate and propagate recombinant baculoviral stocks and to produce recombinant proteins

Gibco Sf21 cells are adapted to serum-free suspension culture in Gibco Sf-900™ II SFM, which saves significant time and expense associated with the adaptation of cultures.

  • Recombinant protein expression from a variety of expression systems
  • Good growth in adherent or suspension culture
  • Documented lineage from a low passage Master Cell Bank
  • Quality and performance testing

Bioproduction, Cell Culture, High-level Protein Production in Insect Cells, Insect Cell Culture, Insect Expression, Insect Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Sf9 and Sf21 Cell Culture, Viral Vaccine Production

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: Liquid nitrogen (vapor phase)
Shipping conditions: Frozen
Cell Line Sf21
Cell Type Insect Cells
Species S. frugiperda
Media Recommendation Sf-900 II SFM (Serum-Free Media)
Product Type Insect Cells
Quantity 1.5 mL
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.