missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ SDF1A Recombinant Human Protein

Product Code. 10120173
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10120173 25 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10120173 Supplier Gibco™ Supplier No. PHC1346

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant SDF1A is a bioactive protein intended for use in cell culture applications

SDF1A is a small cytokine that belongs to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF, or IL-1.

Approved for shipment on Wet or Dry Ice.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Gene ID (Entrez) 6387
Molecular Weight (g/mol) 8 kDa
Purification Method Sequential Chromatography
Quantity 25 μg
Gene Alias SDF-1α
Species Human
Recombinant Recombinant
Content And Storage Store in refrigerator (2–8°C).
Classification Carrier-Free
Endotoxin Level < 0.1 ng/μg
Mass 25 μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Chemokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype SDFs (Stromal Cell-Derived Factors)
Research Category Immunology, Stem Cell Research, Cardiovascular Research, Neurobiology
Product Line Gibco
Purity or Quality Grade 95 %
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.