missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ RANTES Recombinant Human Protein

Product Code. 10079352
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10079352 10 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10079352 Supplier Gibco™ Supplier No. PHC1054

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant RANTES is a bioactive protein intended for use in cell culture applications

RANTES functions as a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. RANTES is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor CCR5 and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor.

Shipping Condition: Dry Ice.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Gene ID (Entrez) 6352
Molecular Weight (g/mol) 7.8 kDa
Purification Method Sequential Chromatography
Quantity 10 μg
Gene Alias RANTES
Species Human
Recombinant Recombinant
Content And Storage Store in refrigerator (2–8°C).
Classification Carrier-Free
Endotoxin Level < 0.1 ng/μg
Mass 10 μg
Shipping Condition Wet Ice
Expression System E. coli
Protein Family Chemokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype Rantes
Research Category Immunology, Stem Cell Research, Oncology
Product Line Gibco
Purity or Quality Grade 95 %
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.