missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS (10X), pH 7.2
GREENER_CHOICE

Product Code. p-4924960
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 15460624

Brand: Gibco™ 70013032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution (see protocol for details).

We offer a variety of Gibco™ PBS formulations for a range of cell culture applications.

This PBS is manufactured as follows:

Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 10 X
For Use With (Application) Mammalian Cell Culture, Chromatin Biology
pH 7.1 to 7.3
Physical Form Liquid
Product Line Gibco
Quantity 500 mL
Solution Type Phosphate Buffered Saline
Sterility Sterile-filtered
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 2800 - 3200 mOsm/kg
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.