missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Opti-MEM™ Reduced Serum Medium, powder

Product Code. 10134472
Change view
Click to view available options
Quantity:
10 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10134472 10 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10134472 Supplier Gibco™ Supplier No. 22600134

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Opti-MEM™ Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change to growth rate or morphology.

Opti-MEM™ Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change to growth rate or morphology. Opti-MEM™ is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Gibco™ Opti-MEM™ modifications for a range of cell culture applications.

This Opti-MEM™ is modified as follows:
With: L-glutamine, Phenol Red

The complete formulation is confidential. For more information, please contact Technical Services.

Gibco™ Opti-MEM™ is a unique media that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%.

Gibco™ Opti-MEM™ uses a sodium bicarbonate buffer system (2.4 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment and filtration at the time of preparation (see protocol for details).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Sp2, AE-1, CHO, BHK-21, and HEK
Cell Type Primary Fibroblasts
Classification Animal Origin
Form Powder
Product Type Opti-MEM I
Sterility Sterile-filtered
With Additives Glutamine, Phenol Red
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco, Opti-MEM
Quantity 10 L
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 30 months from date of manufacture
Show More Show Less

Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.