missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Newborn Calf Serum, New Zealand origin
General stem cell culture media and reagents; sera for specialty research and specific assays
Brand: Gibco™ 16010167
Code : 50
Additional Details : Weight : 0.10000kg
Description
- Sera Category: Secure
- Origin: New Zealand
- Collected from calves 14 days old or less
Cell Culture, Mammalian Cell Culture
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Cattle/Bovine | |
Liquid | |
Newborn Calf Bovine Serum | |
Standard (Sterile-filtered) | |
Sustainable packaging | |
Frozen |
Newborn (20 days or less) | |
New Zealand | |
Bottle | |
100 mL | |
Sterile | |
Gibco™ |