missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Donor Bovine Serum, New Zealand Origin
GREENER_CHOICE

Product Code. 11500516
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11500516 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11500516 Supplier Gibco™ Supplier No. 16030074

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera Category: Secure

Origin: New Zealand.

Endotoxin level: ≤10 EU/ml.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Cattle/Bovine
Age 12 to 36 months
Country of Origin New Zealand
Form Liquid
Format Bottle
Product Type Calf Bovine Serum
Quantity 500 mL
Serum Treatment Sterile-filtered
Sterility Sterile-filtered
Endotoxin Concentration ≤ 10 EU/mL
Product Line Gibco
Shipping Condition Frozen
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.