missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Penicillin-Streptomycin-Neomycin (PSN) Antibiotic Mixture

Product Code. 11580496
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11580496

Brand: Gibco™ 15640055

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Prevents bacterial contamination of cell cultures due to effective combined action against gram-positive and gram-negative bacteria

Penicillin was originally purified from fungus Penicillium and acts by interfering directly with turnover of bacteria cell wall and indirectly by triggering the release of enzymes that alter cell wall. Streptomycin was originally purified from Streptomyces griseus and Neomycin from Streptomyces fradiae. Streptomycin and Neomycin act by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria.

  • Solution contains 5 mg of penicillin, 5 mg of streptomycin, and 10 mg of neomycin per ml in 0.85% saline

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -20°C).
Concentration 100 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Antibiotic
Sterility Sterile-filtered
For Use With (Application) Prevention of Cell Culture Contamination
Quantity 100 mL
Shipping Condition Dry Ice
compliance-icons compliance-icons
Product Identifier
  • Penicillin-Streptomycin-Neomycin (PSN) Antibiotic Mixture
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H360-May damage fertility or the unborn child.
  • H370-Causes damage to organs.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : neomycin sulphate, strepto,ycin sulphate, penicillin G sodium salt

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.