missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MIP-1-β Recombinant Human Protein

Product Code. 10391833
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
1µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10391833 10 μg 1µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10391833 Supplier Gibco™ Supplier No. PHC1034Z

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant MIP-1-beta is a bioactive protein intended for use in cell culture applications

MIP-1-beta (Macrophage inflammatory protein-1β) is involved in a number of biological processes including chemotaxis and is involved in inducing the synthesis of pro-inflammatory cytokines including TNF, IL-6, and IL-1.

Shipping Condition: Dry Ice.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture, Drug Discovery and Development, Target and Lead Identification and Validation, Kinase Biology, Kinase Assay
Gene ID (Entrez) 6351
Molecular Weight (g/mol) 8kDa
Purification Method Sequential Chromatography
Quantity 10 μg
Source Human
Storage Requirements Store at -5° to -30°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Endotoxin Concentration <0.1ng/μg
Gene Alias MIP-1β
Expression System E. coli
Protein Family Chemokines & Receptors
Form Liquid
Protein Subtype MIP (Macrophage Inflammatory Protein) Chemokines
Purity or Quality Grade 95%
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.