missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM, autoclavable, no glutamine, powder

Product Code. 10398102
Click to view available options
Quantity:
10 L
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10398102

Brand: Gibco™ 11700077

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Requires

Supplementation

Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media.

Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This MEM is modified as follows:

With: Phenol Red, Earle's salts

Without: L-glutamine, HEPES, Sodium Bicarbonate

The complete formulation is available.

Gibco MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow's MEM, MEM α, DMEM, and Temin's Modification. To allow autoclaving this medium, choline chloride is substituted with choline bitartrate, phenol red is reduced, and succinic acid is added to prevent precipitation at low pH. This MEM formulation contains Earle's salts for use in a CO2 incubator. This product is made with Earle's salts.

MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

This autoclavable form of Gibco™ cell culture medium requires pH adjustment to 4.1–4.2 before autoclaving, following by sodium bicarbonate and L-glutamine supplementation and final pH adjustment to 7.2–7.4 (see protocol for details).

Please note that the shelf life on delivered products varies. This product has a minimum shelf life of 6 months at time of shipment. For more information regarding product shelf life, including details on specific product lots, please contact Customer Service.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Cell Type Primary Rat Astrocytes
Classification Animal Origin-free
Form Powder
Product Type MEM (Minimum Essential Medium)
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
With Additives Phenol Red, Earle's Balanced Salt Solution (EBSS)
Without Additives No HEPES, No Sodium Bicarbonate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 10 L
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
Show More Show Less

Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For in vitro diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.