Learn More
Gibco™ Human CCL2 (MCP-1) Recombinant Protein
Description
Carrier-Free Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in sterile, distilled water to 0.1-0.5 mg/mL. Apportion the reconstituted protein into working aliquots and store at ≤-20°C. Make any further dilutions of the reconstituted protein in medium or buffered solution containing carrier protein, such as PBS with 0.1% BSA. Storage: Store the lyophilized protein at 2-8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤-20°C (not in a frost-free freezer). Avoid repeated freeze-thaw cycle.
Specifications
Specifications
| Accession Number | P13500 |
| For Use With (Application) | Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 6347 |
| Molecular Weight (g/mol) | 8 kDa |
| Name | Human CCL2 (MCP-1) |
| Purification Method | Purified |
| Quantity | 25 μg |
| Storage Requirements | 4°C |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.