missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Kanamycin Sulfate

Product Code. 11588676
Change view
Click to view available options
Quantity:
5 g
25 g
100 mL
Form:
Liquid
Powder
Unit Size:
100mL
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form unitSize
11588676 25 g Powder 25g
15470594 100 mL Liquid 100mL
11578676 5 g Powder 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11588676 Supplier Gibco™ Supplier No. 11815032

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Water-soluble antibiotic originally purified from bacterium Streptomyces kanamyceticus

  • A water-soluble antibiotic originally purified from the bacterium Streptomyces kanamyceticus
  • Kanamycin acts by binding to the 30S subunit of the bacterial ribosome and inhibiting protein synthesis in susceptible bacteria
  • Gibco™ Kanamycin is effective against a wide variety of gram-negative and some gram-positive bacteria
  • It is used for the prevention of bacterial contamination of cell cultures
  • Provided as a 10 mg/mL stock solution, the recommended working concentration for cell culture is 100 μg/mL
  • Manufactured at a cGMP compliant facility that is registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards
  • Store at -5°C to -20°C

For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Form Powder
Product Type Antibiotic
For Use With (Application) Bacterial Selection
Quantity 25 g
Shipping Condition Ambient Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.