missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Kallikrein 7 Recombinant Human Protein
Recombinant Kallikrein 7 is a bioactive protein intended for use in cell culture applications
POA
Specifications
Purification Method | Sequential Chromatography |
---|---|
Gene Alias | Kallikrein 7 |
Gene ID (Entrez) | KLK7 |
Accession Number | NM_005046 |
Description
KLK7 is one of the Kallikreins, which are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis.Shipping Condition: Wet Ice.
Specifications
Sequential Chromatography | |
Kallikrein 7 | |
KLK7 | |
NM_005046 |
For Research Use Only. Not for use in diagnostic procedures.