missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-8 (CXCL8) (Monocyte-derived) Recombinant Protein

Product Code. 10282833
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
50 μg
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10282833 50 μg Each
10272833 25 μg Each
10477063 100 μg Each
10202553 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10282833 Supplier Gibco™ Supplier No. PHC0885

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant IL8 Monocyte is a bioactive protein intended for use in cell culture applications

Human IL-8 (CXCL8) (Monocyte-derived) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

IL8 is chemotactic for all known types of migratory immune cells. IL8 also inhibits the adhesion of leukocytes to activated endothelial cells.

Shipping Condition: Wet Ice.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Gene ID (Entrez) 3576
Molecular Weight (g/mol) 8.6 kDa
Purification Method Sequential Chromatography
Quantity 50 μg
Gene Alias IL-8 Monocyte
Species Human
Content And Storage Store in refrigerator (2–8°C).
Classification Carrier-Free
Endotoxin Level < 0.1 ng/μg
Mass 50 μg
Shipping Condition Wet Ice
Expression System E. coli
Protein Family Chemokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype Interleukins
Research Category Immunology, Angiogenesis, Respiratory Research, Virology, Stem Cell Research, Inflammation
Product Line Gibco
Purity or Quality Grade 95 %
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.