missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-8 (CXCL8) (Monocyte-derived) Recombinant Protein
Description
Human IL-8 (CXCL8) (Monocyte-derived) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL8 is chemotactic for all known types of migratory immune cells. IL8 also inhibits the adhesion of leukocytes to activated endothelial cells.
Shipping Condition: Wet Ice.
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Gene ID (Entrez) | 3576 |
| Quantity | 25 μg |
| Gene Alias | IL-8 Monocyte |
| Species | Human |
| Content And Storage | Store in refrigerator (2–8°C). |
| Shipping Condition | Wet Ice |
| Expression System | E. coli |
| Protein Family | Chemokines & Receptors |
| Protein Form | Recombinant, Ligand |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?