missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ IL6 Recombinant Mouse Protein
Description
IL6 is a pleiotropic cytokine involved in several cellular processes including inflammation, hematopoeisis, and immune response.
- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of mouse B9 cells
Shipping Condition: Wet Ice.
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Gene ID (Entrez) | 16193 |
| Molecular Weight (g/mol) | ∼20 kDa |
| Purification Method | Sequential Chromatography |
| Quantity | 10 μg |
| Gene Alias | IL-6 |
| Species | Mouse |
| Recombinant | Recombinant |
| Content And Storage | Store in refrigerator (2–8°C). |
| Classification | Carrier-Free |
| Show More |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?