missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ IL33 Recombinant Human Protein
Recombinant IL33 is a bioactive protein intended for use in cell culture applications
POA
Specifications
Purification Method | Sequential Chromatography |
---|---|
Gene Alias | IL-33 |
Gene ID (Entrez) | 90865 |
Accession Number | NM_033439 |
Description
IL33 is a proinflammatory cytokine with structural homology similar with IL-1 family cytokines. IL33 exerts its biological effects by binding to the receptor ST2 and forming a complex with IL-1 Receptor Accessory Protein. Intracellular proteins within in the NFκB and MAP kinase signaling pathways are activated leading to the production of the type 2 cytokines IL-5 from polarized TH2 cells.Shipping Condition: Wet Ice.
Specifications
Sequential Chromatography | |
IL-33 | |
90865 | |
NM_033439 |
For Research Use Only. Not for use in diagnostic procedures.