Learn More
Gibco™ Human IL-21 Recombinant Protein
Description
Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute lyophilized recombinant human IL-21 in sterile, distilled water to a concentration of 0.1-1.0 mg/mL. Further dilutions should be made in low endotoxin medium or a buffered solution containing a carrier protein such as heat inactivated FCS or tissue culture grade BSA. Storage: Store lyophilized recombinant human IL-21 at 2°C to 8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤-20°C. Avoid repeated freeze/thaw cycles.
Specifications
Specifications
| Accession Number | Q9HBE4 |
| For Use With (Application) | Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 59067 |
| Molecular Weight (g/mol) | 15 kDa |
| Name | Human IL-21 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Storage Requirements | 4°C |
| Regulatory Status | RUO |
| Vedi altri risultati |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.