missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IL-2 Recombinant Protein

Product Code. 10662264
Click to view available options
Quantity:
1 mg
10 μg
100 μg
25 μg
Unit Size:
100µg
10µg
1mg
25µg
This item is not returnable. View return policy

Product Code. 10662264

Brand: Gibco™ PMC0025

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant IL-2 is a bioactive protein intended for use in cell culture applications

Mouse IL-2 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

IL-2 causes proliferation of T cells and is a central regulator of immune responses.

Shipping Condition: Wet Ice.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Gene ID (Entrez) 16183
Molecular Weight (g/mol) ∼17 kDa
Purification Method Sequential Chromatography
Quantity 25 μg
Gene Alias IL-2
Species Mouse
Content And Storage Store in refrigerator (2–8°C).
Classification Carrier-Free
Endotoxin Level < 0.1 ng/μg
Mass 25 μg
Shipping Condition Wet Ice
Expression System E. coli
Protein Family Cytokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype Interleukins
Research Category Immunology, Signal Transduction, Stem Cell Research, Angiogenesis, Oncology
Product Line Gibco
Purity or Quality Grade 95 %
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.