missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL2 Recombinant Mouse Protein

Recombinant IL-2 is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMC0024

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10374733

  • £55.17 / Each


Estimated Shipment
31-01-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

IL-2 causes proliferation of T cells and is a central regulator of immune responses.

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture
16183
Sequential Chromatography
10μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
∽17kDa
95%
Mouse
Carrier Protein
Interleukins
IL-2
Cytokines & Receptors
Videos