missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse IL-2 Recombinant Protein
Description
Mouse IL-2 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL-2 causes proliferation of T cells and is a central regulator of immune responses.
Shipping Condition: Wet Ice.
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Gene ID (Entrez) | 16183 |
| Molecular Weight (g/mol) | ∼17 kDa |
| Purification Method | Sequential Chromatography |
| Quantity | 100 μg |
| Gene Alias | IL-2 |
| Species | Mouse |
| Content And Storage | Store in refrigerator (2–8°C). |
| Classification | Carrier-Free |
| Endotoxin Level | < 0.1 ng/μg |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?