missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IL-1 beta Recombinant Protein

Product Code. 10354303
Change view
Click to view available options
Quantity:
10 μg
100 μg
25 μg
5 μg
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10354303 25 μg Each
10324543 100 μg Each
10444943 5 μg Each
10191193 10 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10354303 Supplier Gibco™ Supplier No. PMC0816

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Carrier-Free Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in sterile, distilled water to a concentration of 0.1-1.0 mg/mL. Apportion the reconstituted protein into working aliquots and store at ≤-20°C. Make any further dilutions of the reconstituted protein in low endotoxin medium or buffered solution containing a carrier protein such as heat inactivated FCS or tissue culture grade BSA. Storage: Store the lyophilized protein at 2-8°C or -20°C for long term storage, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤-20°C. Avoid repeated freeze-thaw cycles.

Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P10749
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 16176
Molecular Weight (g/mol) 17 kDa
Name Mouse IL-1 beta
Purification Method Purified
Quantity 25 μg
Storage Requirements 4°C
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/μg
Gene Alias catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1B; IL1B1; IL1beta; IL-1beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1b; interleukin-1beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta
Common Name IL-1 beta
Gene Symbol IL1B
Biological Activity ED50 < 10 pg/mL; determined by the dose-dependent proliferation of murine D10S cells.
Product Type Protein
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Sequence VPIRQLHYRL RDEQQKSLVL SDPYELKALH LNGQNINQQV IFSMSFVQGE PSNDKIPVAL GLKGKNLYLS CVMKDGTPTL QLESVDPKQY PKKKMEKRFV FNKIEVKSKV EFESAEFPNW YISTSQAEHK PVFLGNNSGQ DIIDFTMESV SS
Content And Storage 4°C
Classification Carrier-Free
Activity ED50 < 10 pg/mL; determined by the dose-dependent proliferation of murine D10S cells.
Endotoxin Level <0.1 ng/μg
Shipping Condition Wet Ice
Expression System E. coli
Protein Family Cytokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype Interleukins
Research Category Stem Cell Research, Immunology
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.