missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-1 beta Recombinant Protein

Product Code. 10394403
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
25 μg
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10394403 2 μg Each
10415533 100 μg Each
10111113 1 mg Each
10532635 10 μg Each
10600934 25 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10394403 Supplier Gibco™ Supplier No. PHC0814

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Carrier-Free Reconstitution: We recommend that the vial be briefly centrifuged prior to opening to bring the contents to the bottom. Lyophilized hIL-1 beta should be reconstituted in deionized water to 0.1-1.0 mg/mL to regain full activity. These stock solutions should be apportioned into working aliquots and stored at ≤-20°C. Further dilutions should be made in low endotoxin medium or buffered solution with FBS or tissue culture grade BSA. Storage: Lyophilized hIL-1 beta should be stored at 2°C to 8°C, preferably desiccated. Store reconstituted hIL-1 beta at ≤-20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01584
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 3553
Molecular Weight (g/mol) 17.5 kDa
Name Human IL-1 beta
Purification Method Purified
Quantity 2 μg
Storage Requirements 4°C
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/μg
Gene Alias catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1B; IL1B1; IL1beta; IL-1beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1b; interleukin-1beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta
Common Name IL-1 beta
Gene Symbol IL1B
Biological Activity ED50 = 1.4 - 4 pg/mL; determined by the dose-dependent proliferation of murine D10S cells.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence APVRSLNCTL RDSQQKSLVM SGPYELKALH LQGQDMEQQV VFSMSFVQGE ESNDKIPVAL GLKEKNLYLS CVLKDDKPTL QLESVDPKNY PKKKMEKRFV FNKIEINNKL EFESAQFPNW YISTSQAENM PVFLGGTKGG QDITDFTMQF VSS
Content And Storage 4°C
Classification Carrier-Free
Activity ED50 = 1.4 - 4 pg/mL; determined by the dose-dependent proliferation of murine D10S cells.
Endotoxin Level <0.1 ng/μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Cytokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype Interleukins
Research Category Immunology, Stem Cell Research, Angiogenesis
Product Line Gibco
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.