missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IFN-gamma Recombinant Protein

Product Code. 10009272
Change view
Click to view available options
Quantity:
1 mg
100 μg
15 μg
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10009272 1 mg Each
10552435 100 μg Each
10212593 15 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10009272 Supplier Gibco™ Supplier No. PMC4033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Purity: >95% by SDS-PAGE Endotoxin Concentration: <0.1 ng/mL Biological Activity: ED50 <0.600 ng/mL Molecular Weight: 15 kDa.

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01580
For Use With (Application) Bioactivity
Formulation Tris with no preservative; pH 7.4
Gene ID (Entrez) 15978
Molecular Weight (g/mol) 15 kDa
Name Mouse IFN-gamma
pH Range 7.4
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/mL
Gene Alias BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN γ; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFNγ; Immune interferon; Interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon γ; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferonγ; M-IFN-g; R-IFN-g
Common Name IFN gamma
Gene Symbol IFNG
Biological Activity ED50 < 0.6 ng/mL; determined by a dose-dependent cytostasis assay of mouse WeHi279 cells.
Product Type Protein
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Sequence CYCHGTVIES LESLNNYFNS SGIDVEEKSL FLDIWRNWQK DGDMKILQSQ IISFYLRLFE VLKDNQAISN NISVIESHLI TTFFSNSKAK KDAFMSIAKF EVNNPQVQRQ AFNELIRVVH QLLPESSLRK RKRSR
Content And Storage -80°C
Activity ED50 < 0.6 ng/mL; determined by a dose-dependent cytostasis assay of mouse WeHi279 cells.
Endotoxin Level <0.1 ng/ml
Shipping Condition Dry Ice
Expression System E. coli
Protein Family Cytokines & Receptors
Protein Form Recombinant, Ligand, Mutant
Form Liquid
Protein Subtype Interferons
Research Category Immunology, Virology, Oncology, Signal Transduction
Product Line Gibco
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.