missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ HGH Recombinant Human Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
HGH is a hormone with multiple effects. HGH is a pleiotropic molecule that binds the GH receptor.
- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to get a biologically active protein
- High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by the dose dependent proliferation of monkey 4MBr-5 cells
Shipping Condition: Dry Ice.
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Molecular Weight (g/mol) | 22 kDa |
| Quantity | 10 μg |
| Gene Alias | HGH |
| Species | Human |
| Recombinant | Recombinant |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Classification | Carrier-Free |
| Activity | 2 ng/ml |
| Bioassay | Proliferation of rat lymphoma line Nb2-11 cells |
| Show More |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction