missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HEPES (1 M)

Product Code. 15490594 Shop All Gibco Products
Change view
Click to view available options
Quantity:
20 mL
100 mL
Unit Size:
100mL
20mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490594 20 mL 20mL
12509079 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490594 Supplier Gibco™ Supplier No. 15630106
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gibco HEPES (4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media. It is membrane impermeable, relatively stable, and has limited interference in key processes.

Gibco HEPES (4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media. The addition of 10–25 mM HEPES provides extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator.

Gibco HEPES (N-2-hydroxyethylpiperazine-N-2-ethane sulfonic acid) is a good buffering choice for many cell culture systems because it:

  • Is membrane impermeable
  • Has limited effect on biochemical reactions
  • Is chemically and enzymatically stable
  • Has a very low visible and UV light absorbance

cGMP manufacturing and quality system

For supply chain continuity, we manufacture this product at two separate facilities located in Grand Island, NY, and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements and are certified to the ISO 13485 standard.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zwitterionic Organic Chemical Buffer
CAS 7365-45-9
Concentration 1 M
For Use With (Application) Chromatin Biology
pH 7 to 7.5
Physical Form Liquid
Quantity 20 mL
Solution Type Buffer
Formulation N-2-Hydroxyethylpiperazine-N-2-Ethane Sulfonic Acid
Sterility Sterile
Sterilization Method Sterile-filtered
Molecular Weight (g/mol) 238.3 g/mol
Grade Biochemical
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.