Learn More
Gibco™ HBSS (10X), no calcium, no magnesium, no phenol red
Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
This HBSS is modified as follows:
With: Glucose
Without: Calcium, Magnesium, Phenol Red, Sodium Bicarbonate
The complete formulation is available.
Concentrated forms of Gibco™ HBSS require sodium bicarbonate addition, pH adjustment, and filtration at the time of preparation (see protocol for details).
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | Hanks' Balanced Salt Solution (HBSS) |
| Concentration | 10 X |
| For Use With (Application) | Variety of Cell Culture Applications; Such as Washing Cells Before Dissociation; Transporting Cells or Tissue Samples; Diluting Cells for Counting; and Preparing Reagents |
| pH | 5.8 - 6.1 |
| Physical Form | Liquid |
| Product Line | Gibco |
| Quantity | 500 mL |
| Solution Type | Hank's Balanced Salt |
| Sterility | Sterile-filtered |
| Includes | No Sodium Pyruvate |
| Show More |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.