missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLFNL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531618
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, H1299, HCT116, MCF-7. Predicted reactivity: Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SLFNL1 belongs to the Schlafen family. The function of the SLFNL1 protein remains unknown. There are four different isoforms.
Specifications
| SLFNL1 | |
| Polyclonal | |
| Unconjugated | |
| SLFNL1 | |
| 4933406A14Rik; schlafen like 1; schlafen-like 1; schlafen-like protein 1; SLFNL1; testicular tissue protein Li 173 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 200172 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q499Z3 | |
| SLFNL1 | |
| Recombinant protein encompassing a sequence within the center region of human SLFNL1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction