missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ G-CSF Recombinant Mouse Protein

Product Code. 10151243
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10151243 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10151243 Supplier Gibco™ Supplier No. PMC2031

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant G-CSF is a bioactive protein intended for use in cell culture applications

G-CSF stimulates the proliferation and differentiation of hematopoietic progenitor cells.

Approved for shipment on Wet or Dry Ice.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Molecular Weight (g/mol) 19 kDa
Purification Method Sequential Chromatography
Quantity 100 μg
Gene Alias G-CSF
Species Mouse
Recombinant Recombinant
Content And Storage Store in refrigerator (2–8°C).
Classification Carrier-Free
Endotoxin Level < 0.1 ng/μg
Mass 100 μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Cytokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype GM-CSF, G-CSF, & M-CSF
Research Category Signal Transduction, Oncology, Stem Cell Research, Differentiation, Drug Development
Product Line Gibco
Purity or Quality Grade 95 %
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.