missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human G-CSF Recombinant Protein

Product Code. 10212973
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10212973 25 μg Each
10600344 100 μg Each
10100773 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10212973 Supplier Gibco™ Supplier No. PHC2035

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Carrier-Free Reconstitution: We recommend that the vial be briefly centrifuged prior to opening to bring the contents to the bottom. Lyophilized hG-CSF should be reconstituted in sterile deionized water to 0.1-1.0 mg/mL to regain full activity. These stock solutions should be apportioned into working aliquots and stored at ≤-20°C. Further dilutions should be made in low endotoxin medium or buffered solution with FBS or tissue culture grade BSA. Storage: Lyophilized Hg-CSF should be stored at 2°C to 8°C, preferably desiccated. Store reconstituted hG-CSF at ≤-20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

G-CSF (Granulocyte colony stimulating factor) is a naturally occurring cytokine that stimulates the production and antibacterial function of neutrophils and monocytes. Human G-CSF is an 18.8 kDa protein containing 175 amino acid residues, and a soluble isoform of the G-CSF receptor has been described. The pleotropic cytokine is produced by activated monocytes, macrophages, endothelial cells, fibroblasts, astrocytes, osteoblasts and bone marrow cells. G-CSF has been shown to have specific effects on the proliferation, differentiation and activation of hematopoietic cells. G-CSF is also expressed by various transformed cells such as carcinoma cells and myeloblastic leukemia cells. G-CSF is encoded by two distinct DNA sequences, resulting in a full size, high activity and a shorter, low activity isoform of G-CSF. G-CSF is highly conserved among species and has been shown to exert its biological functions through interaction with its receptor expressed on the surface of hematopoietic progenitors, neutrophilic granulocytes and certain carcinoma cell lines. Clinical use of G-CSF has been approved for several therapeutic applications, treatment of neonatal infections, therapy of acute myocardial infarction, granulocyte transfusion in patients with neutropenia, in severe infections and sepsis, therapy in chronic autoimmune neutropenia, treatment of acute myeloid leukemias, Sweet's syndrome and AIDS. Further, G-CSF has been shown to be a marker protein for different carcinomas such as bladder cancer and dysfunction of the protein has been linked to Kostmann Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09919
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 1440
Molecular Weight (g/mol) 19 kDa
Name Human G-CSF
Purification Method Purified
Quantity 25 μg
Storage Requirements 4°C
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/μg
Gene Alias C17orf33; colony stimulating factor 3; colony stimulating factor 3 (granulocyte); CSF 3; CSF3; CSF3OS; Csfg; filgrastim; GCSF; G-CSF; Granulocyte colony-stimulating factor; granulocyte-colony stimulating factor; gsf3; lenograstim; MGC45931; MGI-IG; Pluripoietin; RP23-395E10.4
Common Name G-CSF
Gene Symbol Csf3
Biological Activity ED50 = 0.05 - 0.2 ng/mL; determined by the dose-dependent proliferation of mouse NFS-60 cells.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Content And Storage 4°C
Classification Carrier-Free
Activity ED50 = 0.05 - 0.2 ng/mL; determined by the dose-dependent proliferation of mouse NFS-60 cells.
Endotoxin Level <0.1 ng/μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Cytokines & Receptors
Protein Form Recombinant, Ligand, Mutant
Form Lyophilized
Protein Subtype GM-CSF, G-CSF, & M-CSF
Research Category Signal Transduction, Oncology, Stem Cell Research, Differentiation, Drug Development
Product Line Gibco
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.