Learn More
Gibco™ FreeStyle™ CHO Expression Medium
Description
FreeStyle CHO Expression Medium is specifically developed to support the growth and transient transfection of CHO-S cells in suspension without adaptation. This medium requires supplementation with 8 mM glutamine before use.
- Formulation without hypoxanthine and thymidine for use with or without dhfr systems
- Ability to support high-density suspension growth and transient transfection of CHO-S cells
- Animal origin-free, protein-free and chemically defined formulation
- Easy use as part of the FreeStyle MAX CHO Expression System for protein production
- Scalability in spinner flasks and bioreactors
Formulation without hypoxanthine and thymidine for use with or without dhfr systems
- Gibco FreeStyle CHO Expression Medium is made without hypoxanthine and thymidine for use in dihydrofolate reductase (dhfr) amplification systems
- Non-dhfr systems require the addition of HT Supplement before use
- Gibco FreeStyle CHO cells in suspension can be cultured directly into Gibco FreeStyle CHO Expression Medium without adaptation
- Cells adapted to other commercially available serum-free media can be subcultured directly into Gibco FreeStyle CHO Expression Medium, usually without any further adaptation
- Cells usually require adaptation from serum-containing formulations
- Cells cultured in Gibco FreeStyle CHO Expression Medium can be transfected using lipid-based transfection reagents, such as FreeStyle MAX reagent
- Gibco FreeStyle CHO Expression Medium is animal origin–free, protein-free, and chemically defined, allowing for easier purification of your protein of interest
- Gibco chemically defined media contain no proteins, hydrolysates, or components of unknown composition
- Protein production using Gibco FreeStyle CHO Expression Medium can be scaled up in spinner flasks or bioreactors
- The appropriate spinner or impeller speed and seeding density should be optimized for each system
- Depending on the impeller design and speed, it may be necessary to supplement Gibco FreeStyle CHO Expression Medium with additional Pluronic™ F-68 to avoid sheer stress in the culture
Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room temperature
Compliance
Gibco FreeStyle CHO Expression Medium is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Specifications
| Product Type | Expression Medium |
| Cell Line | FreeStyle™ CHO-S Cells, CHO |
| Content And Storage | Storage conditions: 2°C to 8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
| Classification | Chemically Defined, Protein-Free, Serum-Free |
| Culture Type | Suspension Cell Culture |
| Endotoxin Level | ≤0.25 EU/mL, ≤ 0.25 EU/mL |
| Glutamine | No Glutamine |
| Phenol Red Indicator | No Phenol Red |
| Serum Level | Serum-Free |
| Product Line | Freestyle, Gibco |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.