Learn More
Gibco™ Mouse FGF-basic (FGF-2/bFGF) Recombinant Protein
Description
Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute lyophilized mouse FGFb in sterile, distilled water to 0.1-0.5 mg/mL. Stock solutions should be apportioned into working aliquots and stored at ≤-20°C. Further dilution should be made in medium or buffered solution containing carrier protein, such as PBS with 0.1% BSA. Storage: Lyophilized mouse FGFb should be stored at 2°C to 8°C, preferably desiccated. Store reconstituted mouse FGFb at ≤-20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.
Specifications
Specifications
| Accession Number | P15655 |
| For Use With (Application) | Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 14173 |
| Molecular Weight (g/mol) | 17 kDa |
| Name | Mouse FGF-basic (FGF-2/bFGF) |
| Purification Method | Purified |
| Quantity | 1 mg |
| Storage Requirements | 4°C |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.