Learn More
Gibco™ Human FGF-basic (FGF-2/bFGF), AOF Recombinant Protein
Human FGF-basic (FGF-2/bFGF), AOF Recombinant Protein
Brand: Gibco™ PHG6015
Code : 50
Additional Details : Weight : 0.05400kg
Description
Human FGF-basic (FGF-2/bFGF), AOF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
FGF Basic is used in studies of angiogenesis, mitogenesis of fibroblasts, neurite outgrowth in PC-12 cells, receptor binding, and tyrosine protein kinase phosphorylation.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by thymidine incorporation into serum-starved Balb/c 3T3 cells
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
50 ug | |
Human | |
Carrier-Free | |
proliferation assay using balb/c 3T3 cells | |
50 μg | |
E. coli | |
Recombinant, Ligand | |
FGFs (Fibroblast Growth Factors) | |
Gibco™ |
17.2 kDa | |
FGF Basic AOF | |
Store in freezer (-5 to -30°C). | |
0.1 ng/ml | |
< 1 EU/ml | |
Wet Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Oncology, Stem Cell Research, Microbiology, Angiogenesis | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.