Learn More
Gibco™ Fetal Bovine Serum, ultra-low IgG, US origin
Description
Gibco Fetal bovine sera include a broad range of specialty sera, earning the trust of researchers with consistent quality and award-winning support that helps meet your unique research requirements. Ultra-low IgG FBS enhances the quality and accuracy of experimental results by minimizing the potential negative effects of IgG (immunoglobulin G). IgG can interfere with immunological and viral assays or experiments involving antibodies, and some cell types are sensitive to the presence of IgG.
- Sera category: Specialty
- Origin: United States
- Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
IgG levels are less than 5 μg/mL, and the BVD titer is low or not detectable.
This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.
Specifications
Specifications
| Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
| Species | Cattle/Bovine |
| Age | Fetal |
| Country of Origin | United States |
| Form | Liquid |
| Format | Bottle |
| Product Type | Fetal Bovine Serum |
| Quantity | 500 mL |
| Serum Treatment | Reduced IgG |
| Shipping Condition | Frozen |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.