Learn More
Gibco™ Fetal Bovine Serum, qualified, heat inactivated, United States
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Value Plus (Performance)
- Origin: United States.
- Endotoxin level: ≤10 EU/mL
- Hemoglobin level: ≤25 mg/dL
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
| Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
| Species | Cattle/Bovine |
| Age | Fetal |
| Country of Origin | United States |
| Form | Liquid |
| Format | Bottle |
| Product Type | Fetal Bovine Serum |
| Quantity | 100 mL |
| Serum Treatment | Heat-inactivated |
| Endotoxin Concentration | ≤10 EU/mL |
| Show More |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.