Learn More
Gibco™ Fetal Bovine Serum, mesenchymal stem cell-Qualified, Australia Origin, Standard (Sterile-Filtered)
Secure sera for cell culture with least viral risk sera from BSE-free regions
Brand: Gibco™ 12664025
Code : NEW
Additional Details : Weight : 0.61000kg
Description
- Sera Category:
- Specially tested for ability to support expansion and clonal enumeration (colony forming unit fibroblast assay) of mesenchymal stem cells
- Endotoxin level: ≤10 EU/mL
- Hemoglobin level: ≤25 mg/dL
- Country of Origin: Australia
Cell Culture, Mammalian Cell Culture, Mesenchymal Stem Cell Culture, Stem Cell Culture, Stem Cell Research
Order Info
Shipping Condition: Dry Ice
Specifications
Blue tongue virus, bovine adenovirus FA, bovine parvovirus FA, bovine respiratory syncytial virus FA, cytopathic agents, electrophoretic pattern, endotoxin (EU/mL, hemadsorbing agents, hemoglobin (ng/dL), mycoplasma, osmolality, rabies FA, reovirus FA, Sp | |
–5 to –20°C | |
Australia | |
Bottle | |
500 mL | |
Ability to support the expansion and clonal enumeration (colony forming unit fibroblast assay) of mesenchymal stem cells | |
Liquid | |
No | |
MS Cell-qualified |
MS Cell-Qualified | |
Fetal Bovine Serum Mesenchymal Stem Cell-Qualified, Australia, 500mL, Liquid, ≤10 EU/mL Endotoxin Concentration, ≤25mg/dL Hemoglobin Concentration, –5 to –20°C Storage Temperature, Fetal, Cattle/Bovine Species, Dry Ice | |
Liquid | |
Fetal Bovine Serum | |
MS Cell-Qualified | |
≤ 10 EU/mL | |
Basic cell culture, specialty research, and specific assays | |
≤25 mg/dL |
For In Vitro Diagnostic Use.