missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Embryonic Stem Cell FBS, Qualified, One Shot™ Format, US Origin
Secure sera for cell culture with least viral risk sera from BSE-free regions
£64.00 - £788.00
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | Fetal |
Species | Cattle/Bovine |
Country of Origin | United States |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11500526
|
Gibco™
16141079 |
500 mL |
£788.00
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946942
|
Gibco™
16141002 |
50 mL |
£64.00
50mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11590516
|
Gibco™
16141061 |
100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Sera Category: Specialty
- Origin: United States
- Specially tested for the ability to sustain undifferentiated cellular morphology of embryonic stem cells
- Endotoxin level: ≤50 EU/mL (levels routinely ≤ 10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
Cell Culture, Embryonic Stem Cell Culture, Mammalian Cell Culture, Stem Cell Culture, Stem Cell Research
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Cattle/Bovine | |
Liquid | |
Fetal Bovine Serum | |
Sterile | |
Frozen |
Fetal | |
United States | |
Bottle | |
Standard (Sterile-filtered) | |
ES Cell-Qualified |
For In Vitro Diagnostic Use.