missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, charcoal stripped, USDA-approved regions

Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays.

Brand:  Gibco™ 12676011

 View more versions of this product

Product Code. 10602304

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera Category: Specialty
  • Serum Treatment: Stripped with charcoal-dextran
  • Recommended Storage: -5°C to -20°C

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
United States or Mexico
Bottle
50 mL
Research
Dry Ice
Fetal
Liquid
Fetal Bovine Serum
Charcoal-stripped
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.