missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, certified, heat inactivated, United States

Product Code. 11523387
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
10 x 50 mL
Format:
Bottle
One Shot
Unit Size:
500mL
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format unitSize
11523387 100 mL Bottle Each
15898937 50 mL One Shot Each
15808947 10 x 50 mL One Shot Each
11533387 500 mL Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11523387 Supplier Gibco™ Supplier No. 10082139

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera Category: Premium (Performance Plus)
  • Origin: United States
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤15 mg/dL

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Cattle/Bovine
Age Fetal
Country of Origin United States
Format Bottle
Quantity 100 mL
Serum Treatment Heat-inactivated
Endotoxin Concentration ≤5 EU/mL
Hemoglobin Concentration ≤15 mg/dL
Purity or Quality Grade Certified
Shipping Condition Frozen
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.