missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS, no calcium, no magnesium
GREENER_CHOICE

Product Code. 10444402
Change view
Click to view available options
Quantity:
500 mL
1000 mL
5 L
10 L
10 x 500 mL
20 L
Packaging:
Bag
Bottle
Unit Size:
1 L
10 L
5 L
500mL
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10444402 1000 mL Bottle 1 L
15326239 500 mL Bottle 500mL
12559069 10 x 500 mL Bottle Pack of 10
11544556 5 L Bag 5 L
11554556 10 L Bag 10 L
11340972 20 L Bag Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10444402 Supplier Gibco™ Supplier No. 14190136

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

This DPBS is modified as follows:

Without: Calcium, Magnesium, Phenol Red

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DPBS
Color Clear
Concentration 1X
For Use With (Application) Mammalian Cell Culture
pH 7.0 to 7.3
Physical Form Liquid
Product Line Gibco
Quantity 1000 mL
Solution Type Dulbecco's Phosphate Buffered Saline
Packaging Bottle
Sterility Sterile-filtered
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 270 - 300 mOsm/kg
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.