Learn More
Gibco™ DMEM, powder, high glucose
Description
Requires
Supplementation
Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This DMEM is modified as follows:
With: High Glucose, L-glutamine, Phenol Red
Without: Sodium Pyruvate, HEPES, Sodium Bicarbonate
The complete formulation is available.
DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate.
DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Cell Line | HeLa, 293, Cos-7, and PC-12 |
| Cell Type | Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells |
| Classification | Animal Origin-free |
| Form | Powder |
| Product Type | DMEM (Dulbecco's Modified Eagle Medium) |
| Serum Level | Standard Serum Supplementation |
| Sterility | Sterile-filtered |
| With Additives | High Glucose, Glutamine, Phenol Red |
| Without Additives | No HEPES, No Sodium Pyruvate, No Sodium Bicarbonate |
| Glucose Concentration | 4500 mg/L |
| Show More |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.