Learn More
Gibco™ DMEM, high glucose, no phosphates

Description
Requires
Supplementation
Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This DMEM is modified as follows:
With: High Glucose, L-glutamine, Phenol Red
Without: Sodium Pyruvate, Phosphate, HEPES
The complete formulation is available.
DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Compliance
Gibco DMEM is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Specifications
| Cell Line | HeLa, 293, Cos-7, and PC-12 |
| Cell Type | Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells |
| Classification | Animal Origin-free |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | DMEM (Dulbecco's Modified Eagle Medium) |
| Serum Level | Standard Serum Supplementation |
| Sterility | Sterile-filtered |
| With Additives | High Glucose, Glutamine, Phenol Red |
| Without Additives | No HEPES, No Sodium Pyruvate, No Phosphate |
| Show More |
In vitro diagnostic. For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.