Learn More
Gibco™ Collagenase, Type I, powder
Description
- Collagenase is a protease that cleaves the bond between a neutral amino acid (X) and glycine in the sequence Pro-X-Gly-Pro, which is found with high frequency in collagen
- It is unique among proteases in its ability to degrade the triple-helical native collagen fibrils commonly found in connective tissues such as skin, tendon, blood vessels, and bone
- Collagenase disaggregation is suitable for the culture of human tumors, mouse kidney, human adult and fetal brain, and many other tissues including epithelium
- Collagenase is relatively gentle, dissociates well at physiological temperature and pH, and requires no mechanical agitation or special equipment
- Gibco™ Collagenase Type I is isolated from Clostridium histolyticum and packaged as a lyophilized, non-sterile powder for research use in cell or tissue dissociation, and organ perfusions
- Compared to other collagenase preparations, Gibco™ Collagenase Type I has average levels of collagenase, caseinase, clostripain, and tryptic activities, and is well-suited for the digestion of fat, adrenal, and liver cells or tissues
- Gibco™ Collagenase Type I activity is guaranteed to be greater than 125 units/mg
- Store at 2°C to 8°C, protected from light
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Storage conditions: 2°C to 8°C. Protect from light. Shipping conditions: Room temperature Shelf life: 24 months from date of manufacture |
| Form | Lyophilized |
| Product Type | Collagenase |
| Sterility | Non-sterile |
| Quantity | 500 mg |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
1. Add 1 mL Hanks' Balanced Salt Solution (HBSS) with calcium and magnesium directly to 1 g vial of Collagenase. Vortex gently to ensure complete dissolution. Transfer to a clean tube.
2. Determine volume of HBSS (with calcium and magnesium) required to bring collagenase solution to 100 U/µL (1000X stock solution). The activity is lot- specific. Rinse vial with this volume of HBSS (with calcium and magnesium), and combine. Filter sterilize 1000X stock solution with a low protein binding filtration unit.
Example: Assuming the lot you have purchased has an activity of 265 U/mg, this lot will have 265000 Units per mL when you reconstitute collagenase into HBSS (with calcium and magnesium) at 1 g/mL. In order to dilute 265000 U/L to 100000 U/mL (= 100 U/µL), you need to dilute the 1 g/mL enzyme solution 2.65 fold.
Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.