missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Trypsin/EDTA Solution (TE)
GREENER_CHOICE

Product Code. 11664862
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11664862 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11664862 Supplier Gibco™ Supplier No. R001100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

With EDTA and phenol red

Made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas. Widely used for cell dissociation, during routine cell culture passaging and primary tissue dissociation due to its digestive strength. Solution formulated as 0.025% trypsin and 0.01% EDTA in Phosphate Buffered Saline (PBS).

Sustainable packaging

Cell Culture, Corneal Epithelial Cell Culture, Fibroblast Culture, Keratinocyte Culture, Large Vessel Endothelial Cell Culture, Mammalian Cell Culture, Mammary Epithelial Cell Culture, Melanocyte Culture, Microvascular Endothelial Cell Culture, Primary Cell Culture, Smooth Muscle Cell Culture

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP compliant facility that is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Chelators EDTA
Content And Storage Storage conditions: -5 to -20°C
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture
Cell Type Mammalian Cells
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile-filtered
With Additives Phenol Red, EDTA
pH 7.4
Product Line Cascade Biologics
Quantity 100 mL
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.