missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Brinster's, no Glutamine, no phenol red
GREENER_CHOICE

Product Code. 11560386
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11560386

Brand: Gibco™ 11126034

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Brinster's medium for ovum culture (BMOC) is a simple chemically defined media containing bovine serum albumin (BSA) developed by Ralph Brinster for in vitro development of mouse oocytes.

Brinster's medium for ovum culture (BMOC) is a simple chemically defined media containing bovine serum albumin (BSA) developed by Ralph Brinster for in vitro development of mouse oocytes. This medium is a variation of the original formulation as it does not contain antibiotics. Brinster's medium has also been used for culture of other mammalian embryos during the early stages of development.

This Brinster's medium is manufactured as follows:

With: Bovine Serum Albumin, Sodium Pyruvate

Without: L-glutamine, HEPES, Phenol Red

The complete formulation is available.

Gibco™ Brinster's medium is unique from other media as it contains no amino acids or vitamins. Brinster's medium is supplemented with bovine serum albumin required for supporting oocyte maturation and the early stages of mammalian embryo development and pyruvate as an essential energy source.

Product Use

For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Brinster's medium uses a sodium bicarbonate buffer system (2.1 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Compliance

Gibco Brinster's medium is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mouse Oocytes
Classification Animal Origin
Concentration 1 X
Form Liquid
Product Type Brinster's Medium
Sterility Sterile-filtered
With Additives Low Glucose, Sodium Pyruvate, Bovine Serum Albumin (BSA)
Without Additives No Glutamine, No HEPES, No Phenol Red
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 100 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.