missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse CXCL13 (BLC/BCA-1) Recombinant Protein

Product Code. 10425903
Click to view available options
Quantity:
20 μg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10425903

Brand: Gibco™ PMC1614

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute in sterile, distilled water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: The lyophilized protein is stable at room temperature for 1 month, but should be kept at -20°C for long term storage. Working aliquots stored with a carrier protein are stable for at least 3 months at -20°C to -80°C. Avoid repeated freeze/thaw cycles.

CXCL13, also known as BCA-1 and Angie, is a small chemokine strongly expressed in the secondary lymphoid organs. It preferentially promotes the migration of B lymphocytes (both B-1 and B-2) to the follicles of the spleen, lymph nodes, and Peyer's patches. It is produced by macrophages, and in mouse the highest constitutive producers include large peritoneal macrophages, Kuppfer cells and lung macrophages (both CD11c+ and CD11c-). The cellular ligand for CXCL13 is CXCR5 also known as BLR-1 (Burkitt's Lymphoma Receptor 1). CXCR5 is expressed on B cells, Tfh cells and, to a lower degree, some memory T cell populations. CXCL13 plays an important role in the development of chronic lymphocytic leukemia (CCL). CXCL13 is also a plasma biomarker for germinal center activity and generation of broadly neutralizing antibodies against HIV.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O55038
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 55985
Molecular Weight (g/mol) 9.5 kDa
Name Mouse CXCL13 (BLC/BCA-1)
Purification Method Purified
Quantity 20 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/μg
Gene Alias 4631412M08Rik; Angie; ANGIE2; B cell-attracting chemokine 1; b lymphocyte chemoattractant; BCA1; BCA-1; B-cell chemoattractant; B-cell-attracting chemokine 1; B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1); BLC; BLR1L; B-lymphocyte chemoattractant; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chem; chemokine (C-X-C motif) ligand 13; chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant); CXC; CXC chemokine BLC; CXC motif chemokine; CXC motif chemokine 13; C-X-C motif chemokine 13; C-X-C motif chemokine ligand 13; CXCL; Cxcl13; SCYB13; small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cell chemoattractant); small inducible cytokine subfamily B (Cys-X-Cys), member 13; small-inducible cytokine B13
Common Name CXCL13
Gene Symbol Cxcl13
Biological Activity Determined by its ability to chemoattract human B cells.
Product Type Protein
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Sequence ILEAHYTNLK CRCSGVISTV VGLNIIDRIQ VTPPGNGCPK TEVVIWTKMK KVICVNPRAK WLQRLLRHVQ SKSLSSTPQA PVSKRRAA
Content And Storage -20°C
Activity Determined by its ability to chemoattract human B cells.
Endotoxin Level <0.1 ng/μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Chemokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype Other Proteins
Research Category Oncology
Product Line Gibco
Purity or Quality Grade ≥98% by SDS-PAGE and HPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.