missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Blasticidin S HCl, powder

Product Code. 11583677
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11583677

Brand: Gibco™ R21001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Bacterial selective antibiotic in concentration range of 50G–100μg/mL

Blasticidin S is a peptidyl nucleoside antibiotic isolated from Streptomyces griseochromogenes. It is a potent inhibitor of protein synthesis in bacteria and eukaryotes, while also active in fungi, nematodes, and tumor cells. Blasticidin S acts by blocking hydrolysis of peptidyl-tRNA induced by release factors and inhibits peptide bond formation. It is used as a selection agent in both mammalian and bacterial cells. The recommended working concentration ranges from 1 to 30 μg/mL depending on the cell line and 25–100 μg/mL for bacterial selection. Cell death occurs rapidly, and blasticidin-resistant stable mammalian cell lines can be generated in less than one week.

  • Blasticidin S acts quickly by inhibiting peptide bond formation by the ribosome
  • Used for prevention of contamination of cell cultures
  • Recommended working concentration ranges from 2 to 2μg/mL depending on the cell line
  • Resistance is conferred by expression of either one of two Blasticidin S deaminase genes: BSD (2) or bsr

Resistance to blasticidin S is conferred by BSR and BSD, isolated from Bacillus cereus K55-S and Aspergillus terreus respectively. The BSR resistance gene encodes blasticidin S deaminase, which catalyzes the conversion of blasticidin S to deaminohydroxyblasticidin S. Deaminohydroxyblasticidin S is a biologically inactive derivative of blasticidin S and does not interact with or inhibit prokaryotic or eukaryotic ribosomes. The BSD resistance gene also encodes a blasticidin S deaminase, which catalyzes a similar reaction to the BSR deaminase.

For bacterial selection purposes, the salt content of the LB medium must remain low (<90 mM) and the pH should not exceed 7.0 to maintain the activity of basticidin S. A kill curve is recommended in order to determine the minimum effective blasticidin S concentration to kill non-resistant cells.

Applications: Cell Culture, Cloning, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 & Sf21 Cell Culture, Transfection, Transformation

Order Info

Shipping conditions: Ambient

TRUSTED_SUSTAINABILITY

Specifications

Color White, off-white
Content And Storage Storage conditions: -5 to -30°C
Shipping conditions: room temperature
Shelf life: Lot Dependent
Cell Type Eukaryotic Cells, Prokaryotic Cells
Concentration 50 to 100 μg/mL
Culture Type Mammalian Cell Culture, Insect Cell Culture
Form Powder
Product Type Antibiotic
Sterility Sterile
With Additives HEPES
For Use With (Application) Bacterial Selection
Product Line Gibco
Quantity 50 mg
Show More Show Less
compliance-icons
Product Identifier
  • blasticidin powder
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H300-Fatal if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : blasticidin S HCl

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.