missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibberellin A95, TRC
Specifications
Specifications
| Chemical Name or Material | Gibberellin A95 |
| CAS | 78259-50-4 |
| Molecular Formula | C19H22O5 |
| Quantity | 100 mg |
| Synonym | 4a,1-(Epoxymethano)-7,9a-methanobenz[a]azulene, gibb-3-ene-1,10-dicarboxylic Acid Deriv., GA95, Gibberellin GA95, NSC 224287, (1R,4aR,4bR,7S,9aS,10S,10aR)-7-Hydroxy-1-methyl-8-methylene-13-oxo-1,4,4b,5,6,7,8,9,10,10a-decahydro-4a,1-(epoxymethano)-7,9a-methanobenzo[a]azulene-10-carboxylic Acid |
| InChI Formula | InChI=1S/C19H22O5/c1-10-8-17-9-18(10,23)7-4-11(17)19-6-3-5-16(2,15(22)24-19)13(19)12(17)14(20)21/h3,6,11-13,23H,1,4-5,7-9H2,2H3,(H,20,21)/t11-,12-,13-,16-,17+,18+,19-/m1/s1 |
| SMILES | [H][C@@]12[C@@](C[C@@]3(O)CC2)(CC3=C)[C@@H](C(O)=O)[C@]4([H])[C@](CC=C[C@]41O5)(C)C5=O |
| IUPAC Name | (1R,4aR,4bR,7S,9aS,10S,10aR)-7-hydroxy-1-methyl-8-methylene-13-oxo-1,2,4b,5,6,7,8,9,10,10a-decahydro-4a,1-(epoxymethano)-7,9a-methanobenzo[a]azulene-10-carboxylic acid |
| Molecular Weight (g/mol) | 330.37 |
| Formula Weight | 330.15 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?